Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004255.3 | NP_004246.2 | 3 | 384 | ||
GenBank mRNA | AI815421.1 | - | 3 | 398 | ||
AK026615.1 | - | 3 | 274 | |||
AK026623.1 | - | 3 | 278 | |||
BC024240.2 | - | 3 | 358 | 4815806 | ||
CR407649.1 | - | 3 | 244 | |||
DQ987236.1 | - | 3 | 244 | |||
DQ987237.1 | - | 3 | 244 | |||
M22760.1 | - | 3 | 263 |