Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001246.3 | NP_001237.1 | 9 | 1984 | ||
NM_203468.2 | NP_982293.1 | 9 | 2053 | |||
XM_011519212.2 | XP_011517514.1 | 8 | 1854 | |||
GenBank mRNA | BC035738.1 | - | 9 | 2047 | 5730586 |