Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170714.1 | NP_001164185.1 | 4 | 1118 | ||
NM_001170715.1 | NP_001164186.1 | 3 | 823 | |||
NM_001170716.1 | NP_001164187.1 | 3 | 780 | |||
NM_001170717.1 | NP_001164188.1 | 3 | 842 | |||
NM_001170718.1 | NP_001164189.1 | 3 | 867 | |||
NM_001170719.1 | NP_001164190.1 | 3 | 957 | |||
NM_001170720.1 | NP_001164191.1 | 4 | 528 | |||
NM_001170721.1 | NP_001164192.1 | 2 | 1055 | |||
NM_014567.3 | NP_055382.2 | 3 | 842 | |||
XM_005256258.4 | XP_005256315.1 | 3 | 1042 | |||
XM_005256259.4 | XP_005256316.1 | 3 | 3954 | |||
XM_005256260.4 | XP_005256317.1 | 3 | 963 | |||
XM_011523475.2 | XP_011521777.1 | 3 | 991 | |||
XM_017023897.1 | XP_016879386.1 | 3 | 1549 | |||
XM_017023898.1 | XP_016879387.1 | 3 | 1549 | |||
GenBank mRNA | AB040024.1 | - | 3 | 837 | ||
AF218451.1 | - | 3 | 820 | |||
AJ242987.2 | - | 3 | 817 | |||
AK027608.1 | - | 3 | 867 | |||
AK074958.1 | - | 2 | 252 | |||
AK124526.1 | - | 3 | 2262 | |||
AK124815.1 | - | 3 | 826 | |||
AK124844.1 | - | 2 | 633 | |||
AK293808.1 | - | 4 | 1118 | |||
AK294513.1 | - | 3 | 873 | |||
AK295809.1 | - | 3 | 752 | |||
AK302617.1 | - | 4 | 528 | |||
AK314722.1 | - | 3 | 842 | |||
AY545071.1 | - | 2 | 696 | |||
BC062556.1 | - | 2 | 697 | 6427808 |