Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORF4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142418.1 | NP_001135890.1 | 5 | 869 | ||
NM_001142419.1 | NP_001135891.1 | 4 | 802 | |||
NM_001142420.1 | NP_001135892.1 | 3 | 649 | |||
NM_001142421.1 | NP_001135893.1 | 4 | 751 | |||
NM_001142422.1 | NP_001135894.1 | 5 | 851 | |||
NM_001142423.1 | NP_001135895.1 | 4 | 769 | |||
NM_001142424.1 | NP_001135896.1 | 5 | 818 | |||
NM_001142425.1 | NP_001135897.1 | 4 | 789 | |||
NM_001142426.1 | NP_001135898.1 | 3 | 667 | |||
NM_001142427.1 | NP_001135899.1 | 4 | 771 | |||
NM_001142428.1 | NP_001135900.1 | 5 | 800 | |||
NM_001142429.1 | NP_001135901.1 | 5 | 838 | |||
NM_001142430.1 | NP_001135902.1 | 4 | 716 | |||
NM_001142431.1 | NP_001135903.1 | 4 | 698 | |||
NM_001142432.1 | NP_001135904.1 | 5 | 820 | |||
NM_012286.2 | NP_036418.1 | 4 | 820 | |||
GenBank mRNA | AF100620.1 | - | 4 | 670 | ||
AF167174.1 | - | 4 | 665 | |||
AK056012.1 | - | 4 | 657 | |||
AK095298.1 | - | 2 | 1582 | |||
AK126115.1 | - | 6 | 645 | |||
AK126427.1 | - | 4 | 1597 | |||
AK223504.1 | - | 4 | 701 | |||
AK302083.1 | - | 5 | 601 | |||
BC056899.1 | - | 5 | 655 | 3905425 | ||
BC093013.1 | - | 5 | 699 | 30528536 | ||
BM471743.1 | - | 4 | 583 | |||
D14812.1 | - | 4 | 670 |