Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008701.2 | NP_001008701.1 | 6 | 1293 | ||
NM_014921.4 | NP_055736.2 | 5 | 1278 | |||
XM_005259818.3 | XP_005259875.1 | 5 | 1334 | |||
XM_011527796.2 | XP_011526098.1 | 6 | 1349 | |||
XM_011527798.2 | XP_011526100.1 | 5 | 1334 | |||
XM_011527801.2 | XP_011526103.1 | 4 | 789 | |||
XM_017026475.1 | XP_016881964.1 | 6 | 1352 | |||
XM_017026476.1 | XP_016881965.1 | 7 | 1462 | |||
XM_017026477.1 | XP_016881966.1 | 6 | 1352 | |||
XM_017026478.1 | XP_016881967.1 | 6 | 1352 | |||
XM_017026479.1 | XP_016881968.1 | 6 | 1352 | |||
XM_017026480.1 | XP_016881969.1 | 3 | 767 | |||
XM_017026481.1 | XP_016881970.1 | 6 | 1356 | |||
GenBank mRNA | AF307079.1 | - | 5 | 1262 | ||
BC028296.1 | - | 5 | 1037 | |||
BC052627.1 | - | 4 | 1019 |
Set Membership: |
G-protein coupled receptor |