Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032279.1 | NP_001027450.1 | 8 | 1051 | ||
NM_005133.2 | NP_005124.1 | 8 | 1055 | |||
GenBank mRNA | AL713740.1 | - | 5 | 2309 | ||
BC052622.1 | - | 8 | 1012 | |||
BC093726.1 | - | 8 | 1051 | 7939571 | ||
BC093728.1 | - | 8 | 1050 | 7939573 | ||
Y13835.2 | - | 8 | 1024 |