Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014924.4 | NP_055739.2 | 5 | 494 | ||
XM_011536563.2 | XP_011534865.1 | 3 | 198 | |||
GenBank mRNA | AK131251.1 | - | 5 | 494 | ||
AK292810.1 | - | 5 | 466 | |||
BC109089.1 | - | 5 | 479 | 40034573 | ||
BC109090.1 | - | 5 | 479 | 40034574 |