Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318810.1 | NP_001305739.1 | 2 | 1092 | ||
NM_001318811.1 | NP_001305740.1 | 2 | 745 | |||
NM_014926.3 | NP_055741.2 | 2 | 930 | |||
GenBank mRNA | AB020655.1 | - | 2 | 759 | ||
AK290024.1 | - | 2 | 1092 | |||
BC114621.1 | - | 2 | 744 | 40036558 |