Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006417.4 | NP_006408.3 | 4 | 673 | ||
NR_135640.1 | 3 | 209 | ||||
NR_135641.1 | 4 | 676 | ||||
XM_005270380.3 | XP_005270437.1 | 4 | 1911 | |||
XM_011540516.2 | XP_011538818.1 | 4 | 1911 | |||
XM_017000083.1 | XP_016855572.1 | 4 | 1911 | |||
GenBank mRNA | AK302527.1 | - | 4 | 634 | ||
AK316278.1 | - | 3 | 166 | |||
AK316476.1 | - | 4 | 631 | |||
AL589497.1 | - | 3 | 237 | |||
BC022870.1 | - | 4 | 638 | 4071404 | ||
DN994910.1 | - | 4 | 650 |