Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 6 | 735 | ||
NM_006433.4 | NP_006424.2 | 5 | 654 | |||
NM_012483.3 | NP_036615.2 | 6 | 898 | |||
XM_005264084.2 | XP_005264141.1 | 5 | 496 | |||
XM_005264085.2 | XP_005264142.1 | 4 | 471 | |||
XM_005264087.2 | XP_005264144.1 | 5 | 723 | |||
GenBank mRNA | AK296106.1 | - | 3 | 499 | ||
AK304728.1 | - | 6 | 668 | |||
BC023576.2 | - | 5 | 529 | 4766109 | ||
X05044.1 | - | 6 | 758 | |||
X54101.1 | - | 5 | 654 |