Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007469.1 | NP_001007470.1 | 4 | 446 | ||
NM_014266.3 | NP_055081.1 | 4 | 449 | |||
XM_017026193.1 | XP_016881682.1 | 4 | 686 | |||
GenBank mRNA | AF072844.1 | - | 4 | 388 | ||
AF122904.1 | - | 4 | 391 | |||
AF172929.1 | - | 4 | 426 | |||
AL050163.1 | - | 4 | 449 | |||
AY359058.1 | - | 4 | 387 | |||
BC035931.1 | - | 4 | 400 | 4701500 | ||
BC046348.1 | - | 4 | 449 | 5229680 | ||
BC065224.1 | - | 4 | 405 | 5762412 |