Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006694.3 | NP_006685.1 | 1 | 788 | ||
GenBank mRNA | AB016488.1 | - | 1 | 497 | ||
AF115850.2 | - | 1 | 486 | |||
AF131797.1 | - | 1 | 369 | |||
AF151056.1 | - | 1 | 344 | |||
AK312106.1 | - | 1 | 470 | |||
BC000499.1 | - | 1 | 510 | 2820861 | ||
BC000996.2 | - | 1 | 437 | 2901221 | ||
BC001363.1 | - | 1 | 475 | 3050800 | ||
BC001667.1 | - | 1 | 440 | 2823348 | ||
BC004239.1 | - | 2 | 297 | 3626047 | ||
BC019277.1 | - | 1 | 431 | 2823348 | ||
BM907938.1 | - | 549 | ||||
BT007285.1 | - | 1 | 65 | |||
CR456985.1 | - | 1 | 65 |