Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017588.2 | NP_060058.1 | 5 | 459 | ||
NM_052821.3 | NP_438172.1 | 4 | 374 | |||
XM_005272163.1 | XP_005272220.1 | 5 | 510 | |||
GenBank mRNA | AJ011376.1 | - | 4 | 374 | ||
AK000552.1 | - | 5 | 425 | |||
AK025810.1 | - | 5 | 419 | |||
BC001635.1 | - | 5 | 433 | 3538255 | ||
BG773451.1 | - | 5 | 465 |