Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031734.3 | NP_001026904.2 | 5 | 536 | ||
GenBank mRNA | AK300568.1 | - | 5 | 529 | ||
BC063460.1 | - | 5 | 520 | 5179971 | ||
DA243025.1 | - | 5 | 536 |