Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 4 | 828 | ||
NM_177974.2 | NP_816929.1 | 4 | 828 | |||
XM_017021880.1 | XP_016877369.1 | 4 | 923 | |||
GenBank mRNA | AK291026.1 | - | 4 | 585 | ||
AK298564.1 | - | 5 | 543 | |||
AL832333.1 | - | 4 | 600 | |||
AL832840.1 | - | 4 | 270 | |||
AY358086.1 | - | 4 | 779 | |||
BC063480.1 | - | 4 | 782 | 5218924 |