Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198075.3 | NP_932341.1 | 14 | 2196 | ||
XM_005252775.2 | XP_005252832.1 | 14 | 3020 | |||
XM_005252776.2 | XP_005252833.1 | 13 | 2843 | |||
XM_006718132.2 | XP_006718195.1 | 14 | 3095 | |||
XM_006718133.2 | XP_006718196.1 | 13 | 1903 | |||
XM_011519875.2 | XP_011518177.1 | 15 | 2435 | |||
XM_011519876.1 | XP_011518178.1 | 15 | 3091 | |||
XM_011519877.2 | XP_011518179.1 | 14 | 2172 | |||
XM_017017167.1 | XP_016872656.1 | 15 | 2510 | |||
XM_017017168.1 | XP_016872657.1 | 15 | 2749 | |||
GenBank mRNA | AK024495.1 | - | 11 | 4086 | ||
AL831961.1 | - | 3 | 562 | |||
BC016439.1 | - | 12 | 1730 | 3447051 | ||
BC035937.1 | - | 14 | 2182 | 4816105 |