Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318870.1 | NP_001305799.1 | 5 | 488 | ||
NM_178336.2 | NP_848026.1 | 5 | 484 | |||
NM_180982.2 | NP_851313.1 | 5 | 481 | |||
NM_181304.2 | NP_851821.1 | 5 | 667 | |||
NM_181305.2 | NP_851822.1 | 4 | 593 | |||
NM_181306.2 | NP_851823.1 | 5 | 492 | |||
NM_181307.2 | NP_851824.1 | 4 | 419 | |||
GenBank mRNA | AK292011.1 | - | 5 | 471 | ||
BG716120.1 | - | 4 | 582 | |||
BX248749.1 | - | 5 | 466 |