Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027158.1 | 4 | 368 | |||
NR_027159.1 | 4 | 691 | ||||
NR_027160.1 | 5 | 814 | ||||
NR_027161.1 | 4 | 810 | ||||
NR_027162.1 | 5 | 867 | ||||
NR_027163.1 | 6 | 601 | ||||
NR_027164.1 | 6 | 548 | ||||
NR_027165.1 | 5 | 412 | ||||
NR_027166.1 | 5 | 425 | ||||
NR_027167.1 | 5 | 491 | ||||
NR_027168.2 | 6 | 517 | ||||
NR_027169.1 | 5 | 544 | ||||
NR_027170.1 | 6 | 469 | ||||
NR_027667.1 | 5 | 735 | ||||
NR_045021.1 | 5 | 404 | ||||
NR_045022.1 | 6 | 543 | ||||
NR_045027.1 | 3 | 634 | ||||
NR_045028.1 | 4 | 678 | ||||
NR_045029.1 | 5 | 499 | ||||
GenBank mRNA | AK098643.1 | - | 5 | 375 | ||
AK307982.1 | - | 5 | 817 | |||
BC012396.1 | - | 3 | 288 | |||
BC019833.1 | - | 4 | 351 | |||
BC027986.1 | - | 5 | 685 | 4940349 | ||
BC040159.1 | - | 5 | 664 | 5539041 | ||
BI333700.1 | - | 4 | 439 | |||
BP353162.1 | - | 5 | 494 | |||
BQ070126.1 | - | 4 | 429 | |||
BU165462.1 | - | 3 | 561 | |||
BU506593.1 | - | 532 | ||||
BU599615.1 | - | 6 | 555 | |||
BX538291.1 | - | 6 | 420 | |||
CB150672.1 | - | 6 | 498 | |||
CD050014.1 | - | 5 | 398 | |||
CD389053.1 | - | 5 | 375 | |||
CF126952.1 | - | 4 | 607 | |||
CN297624.1 | - | 4 | 337 | |||
CR989654.1 | - | 5 | 478 | |||
DA440445.1 | - | 6 | 494 |