Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JOSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270639.1 | NP_001257568.1 | 2 | 347 | ||
NM_001270640.1 | NP_001257569.1 | 2 | 344 | |||
NM_001270641.1 | NP_001257570.1 | 2 | 347 | |||
NM_001270686.1 | NP_001257615.1 | 2 | 190 | |||
NM_138334.3 | NP_612207.1 | 2 | 193 | |||
XM_011526434.2 | XP_011524736.1 | 2 | 187 | |||
GenBank mRNA | AF247787.1 | - | 1 | 122 | ||
AK131052.1 | - | 2 | 212 | |||
BC051380.1 | - | 2 | 151 | |||
BC062416.1 | - | 2 | 170 | 6503623 | ||
BP328576.1 | - | 2 | 190 | |||
HY056543.1 | - | 2 | 348 |