Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHA6 SIRNA Assays ›
Gene Symbol:
PLEKHA6
Gene Name:
pleckstrin homology domain containing A6
Gene Aliases:
PEPP-3, PEPP3
Chromosome Location:
Chr.1: 204218851 - 204378376 on Build GRCh38
UniGene ID:
Hs.253146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22557
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014935.4
NP_055750.2
5, 6
602
XM_005244966.4
XP_005245023.2
5, 6
469
XM_005244967.4
XP_005245024.2
5, 6
468
XM_005244968.4
XP_005245025.2
5, 6
468
XM_006711212.3
XP_006711275.1
5, 6
695
XM_006711213.3
XP_006711276.2
5, 6
469
XM_006711214.3
XP_006711277.2
5, 6
469
XM_006711215.3
XP_006711278.2
5, 6
469
XM_006711217.3
XP_006711280.2
5, 6
469
XM_006711219.3
XP_006711282.2
5, 6
469
XM_006711221.2
XP_006711284.1
5, 6
793
XM_006711222.3
XP_006711285.1
5, 6
778
XM_011509295.2
XP_011507597.1
5, 6
469
XM_011509296.2
XP_011507598.1
5, 6
469
XM_011509297.2
XP_011507599.1
7, 8
1295
XM_011509298.2
XP_011507600.1
5, 6
578
XM_011509299.2
XP_011507601.2
5, 6
584
XM_011509301.2
XP_011507603.1
5, 6
4474
XM_011509304.2
XP_011507606.1
5, 6
423
XM_017000687.1
XP_016856176.1
5, 6
469
XM_017000688.1
XP_016856177.1
5, 6
469
XM_017000689.1
XP_016856178.1
5, 6
469
XM_017000690.1
XP_016856179.1
5, 6
4474
GenBank mRNA
AB023186.1
-
5, 6
589
BC152475.1
-
5, 6
589
8862612
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline