Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145047.4 | NP_659484.4 | 1 | 207 | ||
NM_206837.2 | NP_996668.1 | 1 | 207 | |||
XM_005270461.1 | XP_005270518.1 | 1 | 207 | |||
XM_005270462.1 | XP_005270519.1 | 1 | 207 | |||
XM_005270463.3 | XP_005270520.1 | 1 | 207 | |||
XM_011540680.2 | XP_011538982.1 | 1 | 174 | |||
XM_011540681.2 | XP_011538983.1 | 1 | 55 | |||
XM_017000303.1 | XP_016855792.1 | 1 | 143 | |||
XM_017000304.1 | XP_016855793.1 | 1 | 143 | |||
GenBank mRNA | AB079075.1 | - | 1 | 91 | ||
AK098541.1 | - | 1 | 173 | |||
BU854079.1 | - | 168 | ||||
DA297022.1 | - | 1 | 207 |