Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037814.1 | NP_001032903.1 | 4 | 878 | ||
XM_011546116.2 | XP_011544418.1 | 4 | 937 | |||
XM_011546117.1 | XP_011544419.1 | 2 | 168 | |||
XM_017028575.1 | XP_016884064.1 | 4 | 930 | |||
GenBank mRNA | AB076978.1 | - | 4 | 878 |