Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294347.1 | NP_001281276.1 | 4 | 2188 | ||
NM_005202.3 | NP_005193.1 | 4 | 2333 | |||
XM_005270477.3 | XP_005270534.1 | 4 | 3033 | |||
GenBank mRNA | AK074129.1 | - | 2 | 2251 |