Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001851.4 | NP_001842.3 | 35, 36 | 2470 | ||
NM_078485.3 | NP_511040.2 | 29, 30 | 1738 | |||
XM_011535429.2 | XP_011533731.1 | 36, 37 | 2500 | |||
XM_011535430.2 | XP_011533732.1 | 30, 31 | 1766 | |||
XM_017010246.1 | XP_016865735.1 | 33, 34 | 1857 | |||
XM_017010247.1 | XP_016865736.1 | 27, 28 | 1713 | |||
GenBank mRNA | AK097582.1 | - | 7, 8 | 1016 | ||
AK125738.1 | - | 24, 25 | 1331 | |||
BC063646.1 | - | 29, 30 | 1712 | 4914907 | ||
X54412.1 | - | 35, 36 | 2442 |