Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 4 | 375 | ||
NM_145261.3 | NP_660304.1 | 4 | 363 | |||
NR_033721.1 | 3 | 311 | ||||
GenBank mRNA | AI268262.1 | - | 4 | 362 | ||
AK309019.1 | - | 2 | 1264 | |||
AK311765.1 | - | 4 | 291 | |||
BC009702.1 | - | 4 | 315 | 3892297 | ||
BC073989.1 | - | 4 | 308 | 5760865 | ||
BE773017.1 | - | 4 | 330 | |||
BF666599.1 | - | 4 | 325 | |||
BG492744.1 | - | 3 | 247 | |||
CB054787.1 | - | 1 | 79 | |||
CB159371.1 | - | 4 | 375 | |||
KU178861.1 | - | 3 | 191 |