Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014446.2 | NP_001014446.1 | 4, 5 | 443 | ||
NM_001286773.1 | NP_001273702.1 | 4, 5 | 466 | |||
NM_001286774.1 | NP_001273703.1 | 3, 4 | 346 | |||
NM_152398.3 | NP_689611.1 | 4, 5 | 443 | |||
GenBank mRNA | AK298305.1 | - | 4, 5 | 412 | ||
AY472117.1 | - | 4, 5 | 445 | |||
BC032808.1 | - | 4, 5 | 414 | 4454708 | ||
BI546814.1 | - | 4, 5 | 448 | |||
BM782144.1 | - | 4, 5 | 413 | |||
BU508058.1 | - | 3, 4 | 234 | |||
CR749310.1 | - | 4, 5 | 388 | |||
W68799.1 | - | 4, 5 | 395 |