Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM173B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258388.1 | NP_001245317.1 | 2 | 83 | ||
NM_001258389.1 | NP_001245318.1 | 2 | 83 | |||
NM_199133.3 | NP_954584.2 | 2 | 83 | |||
NR_047668.1 | 2 | 83 | ||||
NR_047669.1 | 2 | 83 | ||||
NR_047670.1 | 3 | 150 | ||||
XM_011513964.1 | XP_011512266.1 | 2 | 70 | |||
GenBank mRNA | AK055604.1 | - | 3 | 138 | ||
AK300042.1 | - | 2 | 60 | |||
AK302012.1 | - | 2 | 72 | |||
AK316317.1 | - | 2 | 68 | |||
BC063406.1 | - | 2 | 51 | 5190223 | ||
BG611454.1 | - | 2 | 63 | |||
DA523490.1 | - | 2 | 62 | |||
DA843869.1 | - | 2 | 59 | |||
DW420473.1 | - | 2 | 79 |