Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138811.3 | NP_620166.3 | 10 | 1980 | ||
XM_005249619.3 | XP_005249676.1 | 10 | 1829 | |||
XM_005249620.3 | XP_005249677.1 | 11 | 2360 | |||
XM_011515125.2 | XP_011513427.1 | 6 | 871 | |||
GenBank mRNA | AK097248.1 | - | 11 | 1701 | ||
AK098189.1 | - | 5 | 891 | |||
BC015397.1 | - | 3 | 710 | |||
BC043269.2 | - | 10 | 2083 | 5297014 | ||
BC060879.1 | - | 10 | 1893 | 5298874 | ||
BX640851.1 | - | 7 | 1050 |