Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM92A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145269.4 | NP_660312.2 | 6, 7 | 680 | ||
NR_104267.1 | 6, 7 | 680 | ||||
NR_104268.1 | 6, 7 | 680 | ||||
XM_005250787.2 | XP_005250844.1 | 6, 7 | 550 | |||
GenBank mRNA | AK096298.1 | - | 6, 7 | 574 | ||
BC014598.2 | - | 6, 7 | 680 | |||
BC108707.1 | - | 6, 7 | 642 | |||
CR627475.1 | - | 6, 7 | 1719 | |||
DQ327716.1 | - | 6, 7 | 594 |