Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205545.2 | NP_991108.1 | 3 | 313 | ||
GenBank mRNA | AB593150.1 | - | 3 | 286 | ||
AK290331.1 | - | 3 | 292 | |||
AY358432.1 | - | 3 | 313 | |||
BC119019.1 | - | 3 | 309 | 40108704 | ||
BC119812.1 | - | 3 | 309 | 40108706 | ||
DA449804.1 | - | 3 | 292 |