Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024708.3 | NP_078984.2 | 5 | 1465 | ||
NM_198243.2 | NP_937886.1 | 5 | 1465 | |||
GenBank mRNA | AF451994.1 | - | 5 | 1385 | ||
AK026204.1 | - | 5 | 1385 | |||
AK222958.1 | - | 5 | 1385 | |||
AK222974.1 | - | 5 | 1385 | |||
AK289860.1 | - | 5 | 1418 | |||
BC063581.1 | - | 5 | 1406 | 4370796 |