Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017961.4 | NP_001017961.1 | 1 | 724 | ||
NM_001320302.1 | NP_001307231.1 | 1 | 724 | |||
NR_135199.1 | 1 | 724 | ||||
XM_017000417.1 | XP_016855906.1 | 1 | 724 | |||
XM_017000418.1 | XP_016855907.1 | 1 | 275 | |||
GenBank mRNA | AB593134.1 | - | 1 | 724 | ||
AK299946.1 | - | 1 | 254 | |||
BC114214.1 | - | 2 | 807 | 5588830 |