Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152516.3 | NP_689729.1 | 1, 2 | 224 | ||
XM_011532558.2 | XP_011530860.1 | 1, 2 | 222 | |||
GenBank mRNA | AB178811.1 | - | 1, 2 | 208 | ||
AK223607.1 | - | 1, 2 | 213 | |||
AK294203.1 | - | 1, 2 | 192 | |||
BC009266.2 | - | 1, 2 | 177 | |||
BC022046.1 | - | 1, 2 | 196 | 4046433 | ||
D85433.1 | - | 1, 2 | 124 | |||
HY001060.1 | - | 1, 2 | 224 |