Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 14 | 1748 | ||
NM_001303257.2 | NP_001290186.1 | 14 | 1748 | |||
NM_014941.3 | NP_055756.1 | 15 | 2614 | |||
XM_011530004.1 | XP_011528306.1 | 14 | 1973 | |||
XM_017028667.1 | XP_016884156.1 | 14 | 1973 | |||
XM_017028668.1 | XP_016884157.1 | 15 | 2068 | |||
GenBank mRNA | AB020659.1 | - | 15 | 2528 | ||
BC019257.1 | - | 13 | 1275 | 3503651 | ||
BC136782.1 | - | 15 | 1473 | 9020772 | ||
CR456469.1 | - | 13 | 1274 |