Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130899.2 | NP_570969.2 | 1 | 693 | ||
GenBank mRNA | AF465821.1 | - | 1 | 669 | ||
AK057543.1 | - | 1 | 689 | |||
AY973323.1 | - | 1 | 593 | |||
BC022035.1 | - | 1 | 701 | 4826350 | ||
BC025397.1 | - | 1 | 694 | 4827541 | ||
BC025409.1 | - | 1 | 694 | 4825198 | ||
BC025998.1 | - | 1 | 694 | 4836822 |