Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225769
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
15
2126
NM_152586.3
NP_689799.3
15
2224
NR_135249.1
18
2927
NR_135250.1
13
1922
XM_005269582.2
XP_005269639.1
14
5302
XM_011539368.2
XP_011537670.1
15
5368
XM_017015759.1
XP_016871248.1
17
4797
XM_017015760.1
XP_016871249.1
17
3699
XM_017015761.1
XP_016871250.1
17
4537
XM_017015762.1
XP_016871251.1
17
3707
XM_017015763.1
XP_016871252.1
17
3001
XM_017015764.1
XP_016871253.1
17
3064
XM_017015765.1
XP_016871254.1
18
3317
XM_017015766.1
XP_016871255.1
17
4923
XM_017015767.1
XP_016871256.1
17
4582
XM_017015768.1
XP_016871257.1
17
2753
XM_017015769.1
XP_016871258.1
17
2361
XM_017015770.1
XP_016871259.1
17
3022
XM_017015771.1
XP_016871260.1
16
5539
XM_017015772.1
XP_016871261.1
16
5545
XM_017015773.1
XP_016871262.1
15
5479
XM_017015774.1
XP_016871263.1
16
4465
XM_017015775.1
XP_016871264.1
17
4343
XM_017015776.1
XP_016871265.1
16
4848
XM_017015777.1
XP_016871266.1
16
4507
XM_017015778.1
XP_016871267.1
16
5545
XM_017015779.1
XP_016871268.1
16
5545
XM_017015780.1
XP_016871269.1
16
5545
XM_017015781.1
XP_016871270.1
15
5374
XM_017015782.1
XP_016871271.1
17
3245
XM_017015783.1
XP_016871272.1
16
4172
XM_017015784.1
XP_016871273.1
15
2118
XM_017015785.1
XP_016871274.1
16
5545
XM_017015786.1
XP_016871275.1
16
2735
GenBank mRNA
AK126497.1
-
5
895
AK127213.1
-
3
355
AK127267.1
-
13
1922
BC130633.1
-
2
124
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline