Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173526.3 | NP_775797.2 | 6 | 1315 | ||
XM_011536500.1 | XP_011534802.1 | 6 | 1279 | |||
XM_011536502.1 | XP_011534804.1 | 6 | 1214 | |||
XM_011536503.2 | XP_011534805.1 | 5 | 2256 | |||
GenBank mRNA | AK093449.1 | - | 6 | 1279 |