Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144980.3 | NP_659417.2 | 7 | 1325 | ||
XM_005266838.2 | XP_005266895.1 | 7 | 1350 | |||
XM_011535509.2 | XP_011533811.1 | 7 | 1512 | |||
XM_011535510.2 | XP_011533812.1 | 6 | 1452 | |||
XM_011535511.2 | XP_011533813.1 | 7 | 1512 | |||
XM_011535512.2 | XP_011533814.1 | 7 | 1375 | |||
XM_011535513.2 | XP_011533815.1 | 7 | 1778 | |||
XM_017010323.1 | XP_016865812.1 | 7 | 1512 | |||
XM_017010324.1 | XP_016865813.1 | 7 | 1987 | |||
GenBank mRNA | AK302358.1 | - | 7 | 1780 | ||
BC026278.1 | - | 7 | 1312 | 4703212 |