Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288743.1 | NP_001275672.1 | 5 | 387 | ||
XM_011539378.2 | XP_011537680.1 | 5 | 567 | |||
XM_011539379.1 | XP_011537681.1 | 5 | 567 | |||
XM_011539380.1 | XP_011537682.1 | 5 | 567 | |||
XM_017015792.1 | XP_016871281.1 | 5 | 567 | |||
GenBank mRNA | BC037922.2 | - | 5 | 378 | ||
BC042857.1 | - | 5 | 378 | |||
DQ884399.1 | - | 5 | 323 |