Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037325.2 | NP_001032402.1 | 3 | 561 | ||
NM_001199165.2 | NP_001186094.1 | 23 | 2692 | |||
NM_001302891.1 | NP_001289820.1 | 21 | 2642 | |||
XM_005257119.4 | XP_005257176.1 | 23 | 2786 | |||
XM_005257125.3 | XP_005257182.1 | 15 | 1776 | |||
XM_005257126.4 | XP_005257183.1 | 12 | 1441 | |||
XM_006721740.2 | XP_006721803.1 | 23 | 2658 | |||
XM_011524461.2 | XP_011522763.1 | 23 | 2799 | |||
XM_011524462.2 | XP_011522764.1 | 23 | 2788 | |||
XM_011524463.2 | XP_011522765.1 | 23 | 2788 | |||
XM_011524465.2 | XP_011522767.1 | 20 | 2115 | |||
XM_011524466.2 | XP_011522768.1 | 13 | 1594 | |||
XM_011524467.2 | XP_011522769.1 | 12 | 1436 | |||
XM_017024302.1 | XP_016879791.1 | 21 | 2529 | |||
GenBank mRNA | AF458591.1 | - | 13 | 1599 | ||
AK097612.1 | - | 5 | 660 | |||
AK299075.1 | - | 23 | 2643 | |||
BC029524.1 | - | 3 | 561 | |||
BC131805.1 | - | 21 | 2470 | 40119069 | ||
BX097629.1 | - | 3 | 239 |