Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271006.1 | NP_001257935.1 | 5 | 370 | ||
NM_001271007.1 | NP_001257936.1 | 4 | 301 | |||
NM_144998.3 | NP_659435.2 | 4 | 316 | |||
XM_005256339.2 | XP_005256396.1 | 5 | 522 | |||
XM_017024326.1 | XP_016879815.1 | 3 | 723 | |||
XM_017024327.1 | XP_016879816.1 | 4 | 595 | |||
XM_017024328.1 | XP_016879817.1 | 4 | 464 | |||
XM_017024329.1 | XP_016879818.1 | 3 | 410 | |||
GenBank mRNA | BC009571.1 | - | 4 | 285 | 4333630 | |
BC011610.1 | - | 4 | 252 | |||
BI837753.1 | - | 4 | 284 | |||
CB110145.1 | - | 5 | 379 | |||
HY110543.1 | - | 5 | 360 |