Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018941.3 | NP_061764.2 | 3 | 1038 | ||
XM_005266021.3 | XP_005266078.1 | 3 | 982 | |||
XM_005266022.1 | XP_005266079.1 | 3 | 1161 | |||
XM_005266023.1 | XP_005266080.1 | 3 | 879 | |||
XM_011534745.1 | XP_011533047.1 | 3 | 983 | |||
XM_011534746.2 | XP_011533048.1 | 3 | 880 | |||
GenBank mRNA | AF123757.1 | - | 3 | 967 | ||
AF123758.1 | - | 3 | 967 | |||
AF123759.1 | - | 3 | 967 | |||
AF123760.1 | - | 3 | 967 | |||
AF123761.1 | - | 3 | 967 | |||
AK312486.1 | - | 3 | 981 | |||
BC007725.2 | - | 3 | 892 | 4303103 | ||
BT007049.1 | - | 2 | 733 |