Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 13 | 1193 | ||
NM_001286835.1 | NP_001273764.1 | 17 | 2445 | |||
XM_005272748.1 | XP_005272805.1 | 15 | 1297 | |||
XM_005272749.1 | XP_005272806.1 | 14 | 1240 | |||
XM_005272750.1 | XP_005272807.1 | 14 | 1237 | |||
XM_005272751.1 | XP_005272808.1 | 14 | 1210 | |||
XM_006716914.1 | XP_006716977.1 | 14 | 1167 | |||
XM_017014455.1 | XP_016869944.1 | 13 | 1080 | |||
XM_017014456.1 | XP_016869945.1 | 16 | 1228 | |||
XM_017014457.1 | XP_016869946.1 | 15 | 1141 | |||
XM_017014459.1 | XP_016869948.1 | 17 | 1476 | |||
XM_017014461.1 | XP_016869950.1 | 16 | 1389 | |||
GenBank mRNA | AK097639.1 | - | 15 | 1830 | ||
BC043420.1 | - | 3 | 2605 | |||
BC067803.1 | - | 17 | 2383 | 5294979 | ||
BC082271.1 | - | 13 | 1087 | |||
BC141856.1 | - | 16 | 1363 | |||
BM711778.1 | - | 6 | 455 |