Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040274.2 | NP_001035364.2 | 23 | 2237 | ||
GenBank mRNA | AK128130.1 | - | 23 | 2291 | ||
AM293674.1 | - | 23 | 1956 | |||
BC012225.1 | - | 13 | 1134 |