Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 22 | 2827 | ||
NM_001278438.1 | NP_001265367.1 | 22 | 2870 | |||
NM_014923.4 | NP_055738.3 | 20 | 2451 | |||
NR_103528.1 | 22 | 2859 | ||||
XM_011534997.2 | XP_011533299.1 | 21 | 2541 | |||
XM_017020440.1 | XP_016875929.1 | 22 | 2712 | |||
XM_017020441.1 | XP_016875930.1 | 24 | 3333 | |||
XM_017020442.1 | XP_016875931.1 | 23 | 2865 | |||
XM_017020443.1 | XP_016875932.1 | 22 | 3185 | |||
GenBank mRNA | AB023187.2 | - | 20 | 2381 | ||
AJ749706.1 | - | 22 | 2829 | |||
AJ749707.1 | - | 20 | 2442 | |||
AK302415.1 | - | 20 | 2451 | |||
BC132812.1 | - | 22 | 2594 | 40146834 | ||
BC136617.1 | - | 22 | 2595 | 9020607 | ||
BC144301.1 | - | 22 | 2595 | 9052821 | ||
BX640739.1 | - | 22 | 2730 | |||
BX648141.1 | - | 22 | 2817 | |||
BX648706.1 | - | 24 | 2675 |