Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040402.2 | NP_001035492.1 | 7 | 726 | ||
NM_001287755.1 | NP_001274684.1 | 7 | 622 | |||
NM_001287757.1 | NP_001274686.1 | 8 | 889 | |||
NM_015115.3 | NP_055930.2 | 7 | 726 | |||
XM_005265731.3 | XP_005265788.1 | 7 | 616 | |||
XM_011534379.2 | XP_011532681.1 | 7 | 1149 | |||
XM_011534380.1 | XP_011532682.1 | 7 | 485 | |||
XM_017007912.1 | XP_016863401.1 | 7 | 579 | |||
XM_017007913.1 | XP_016863402.1 | 6 | 1064 | |||
GenBank mRNA | AK027051.1 | - | 7 | 545 | ||
AK124346.1 | - | 6 | 1294 | |||
AK294894.1 | - | 7 | 622 | |||
AK294896.1 | - | 8 | 736 | |||
BC053897.1 | - | 7 | 592 | 6173607 | ||
BX537944.1 | - | 7 | 553 | |||
D87466.1 | - | 7 | 536 |