Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 10 | 1340 | ||
NM_001281711.1 | NP_001268640.1 | 10 | 1301 | |||
NM_001281712.1 | NP_001268641.1 | 9 | 1120 | |||
NM_015341.4 | NP_056156.2 | 10 | 1373 | |||
XM_005263908.3 | XP_005263965.1 | 10 | 1313 | |||
XM_006712388.3 | XP_006712451.1 | 10 | 1313 | |||
GenBank mRNA | AK291053.1 | - | 10 | 1314 | ||
AK299248.1 | - | 10 | 1242 | |||
AK303725.1 | - | 9 | 1065 | |||
AK308050.1 | - | 10 | 1280 | |||
AK310649.1 | - | 10 | 1241 | |||
BC024211.2 | - | 10 | 1327 | 3504782 | ||
D38553.1 | - | 10 | 1303 |