Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 11 | 1679 | ||
XM_011518446.2 | XP_011516748.1 | 11 | 1694 | |||
XM_011518447.1 | XP_011516749.1 | 10 | 1655 | |||
XM_011518449.1 | XP_011516751.1 | 10 | 1562 | |||
XM_011518451.1 | XP_011516753.1 | 9 | 1550 | |||
XM_011518452.1 | XP_011516754.1 | 10 | 1445 | |||
XM_017014571.1 | XP_016870060.1 | 10 | 1652 | |||
XM_017014572.1 | XP_016870061.1 | 9 | 1421 | |||
GenBank mRNA | AF153415.1 | - | 11 | 1679 | ||
AK172815.1 | - | 5 | 424 | |||
AK299952.1 | - | 11 | 1661 | |||
AL556162.3 | - | 5 | 390 | |||
AL833960.1 | - | 9 | 1070 | |||
BC014248.2 | - | 1 | 30 |