Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289910.1 | NP_001276839.1 | 4 | 343 | ||
NM_001290114.1 | NP_001277043.1 | 2 | 318 | |||
NM_002168.3 | NP_002159.2 | 4 | 576 | |||
GenBank mRNA | AK222916.1 | - | 4 | 490 | ||
AK294148.1 | - | 4 | 343 | |||
AK299987.1 | - | 2 | 232 | |||
AK312627.1 | - | 4 | 540 | |||
AK316388.1 | - | 4 | 417 | |||
BC009244.2 | - | 4 | 478 | 2959540 | ||
BC071828.1 | - | 4 | 477 | 6090792 | ||
X69433.1 | - | 4 | 498 |