Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014949.3 | NP_055764.2 | 4 | 499 | ||
GenBank mRNA | AB020714.1 | - | 4 | 456 | ||
AK225714.1 | - | 4 | 470 | |||
AK289902.1 | - | 4 | 442 | |||
AY112680.1 | - | 4 | 430 | |||
AY112681.1 | - | 4 | 430 | |||
AY112682.1 | - | 4 | 430 | |||
BC027182.1 | - | 4 | 455 | |||
BC062637.1 | - | 4 | 454 | 5497581 | ||
BI560023.1 | - | 4 | 509 | |||
KU178555.1 | - | 4 | 430 | |||
KU178556.1 | - | 4 | 430 |